Learn More
BRAND™ PTFE Watch Glass
PTFE construction. Ideal for covering PTFE beakers.
Brand:  BRAND™ 150912
20.95 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10718361
  • 21.50 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • PTFE construction
  • Excellent chemical resistance
  • Ideal for covering PTFE beakers
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Flat
Round
80 mm
PTFE
Watch Glass