missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Puerckhauer soil sampler

Buerkle™ Puerckhauer soil sampler for heavy and stony soils.

Brand:  Buerkle™ 5350-5012

 View more versions of this product

Product Code. 10618898

  • 708.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Very robust Puerckhauer-sampler for heavy and stony soils. Model 5350-5018 has a conically-tapered 60 cm tip which makes it much easier to extract the driven-in sampler. Ideal for N-min sampling0–90 cm.
  • Depth graduation 10 cm divisions
  • Attachable handle, with non-slip plastic grips
  • Reinforced impact head
  • Stainless steel
Specifications

Specifications

Puerckhauer Soil Sampler
Attachable handle, with non-slip Plastic grips
Yes
2400 g
20 mm
Stainless Steel
T-shape
For Heavy and Stony Soils, 0 to 90 cm N-Min Sampling
20 mm
81 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers