missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pumice Stone, Granular, 4 to 6mm, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Poly Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Specifications
Specifications
Packaging | Poly Bottle |
Odor | Odorless |
Color | Gray |
Quantity | 1 kg |
UN Number | NONH for all modes of transport |
Chemical Name or Material | Pumice stone |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Pumice Stone, Granular, 4 to 6mm, Honeywell Fluka™ >
Spot an opportunity for improvement?Share a Content Correction