missing translation for 'onlineSavingsMsg'
Learn More

Masterflex™ L/S™ 2-Channel, 6-Roller Cartridge Pump Head

Synchronous, low-pulsation flow from multiple channels

Brand:  Masterflex™ 07519-10

Product Code. 11865252

  • 610.00 € / Each
Estimated Shipment: 28-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Cartridges snap in and out for fast tubing changes—no need to remove pump head from the drive
  • Improved retainer design holds tubing securely in place
  • Flow rate 0.0008 to 350 mL/min with L/S tubing—flow rate depends on drive rpm, cartridge, and tubing size
  • Accepts up to 2 large cartridges
  • Require drives with a minimum of 1⁄10 hp and a maximum of 250 rpm
Specifications

Specifications

100 x 111 x 102 mm
Polysulfone (PSF)
6
100 mm
350 mL/min (max.)
2 Large
102 mm
111 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.