Learn More
BINDER™ Pure Aqua Service
Pure Aqua Service system for preparation or complete desalination of tap water. For use with series KBWF, KBF, KBF P, KBF LQC, KMF, KBF-S, MKF, and MKFT.
Brand: BINDER™ 8012-0759
This item is not returnable.
View return policy
Description
- Suitable for: KBF-S 720, MKF 240, KBF LQC 240, KBF P 720, KBF-S 240, KBF P 240, KBWF 720, MKFT 720, MKFT 240, MKFT 115, MKF 56, MKF 720, KBF LQC 720, KBF 720, KBWF 240, MKF 115, KMF 720, KMF 240, KMF 115, KBF 1020, KBF 240, KBF 115
Specifications
BINDER PURE AQUA SERVICE - System for preparation or complete desalination of tap water, complete set containing PURE AQUA 300 single-use cartridge, measuring device, and all necessary connecting parts | |
BINDER Chambers Series KBWF, KBF, KBF P, KBF LQC, KMF, KBF-S, MKF, MKFT | |
Plastic | |
Water Supply |