missing translation for 'onlineSavingsMsg'
Learn More
Learn More
uvex Pure-Fit Safety Glasses

Glasses designed to be recycled. Pure-Fit can be manufactured fully automatically from just one material without the need for additional assembly.
5.50 € - 7.55 €
Specifications
Lens Tint | Clear |
---|---|
Product Type | Ergonomic Safety Glasses |
Frame Color | Transparent |
Material | Polycarbonate |
Disposable | Reusable |
Product Code | Brand | Lens Coating | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Lens Coating | Price | Quantity & Availability | ||||||
17523085
![]() |
uvex
9145014 |
Uncoated |
5.50 €
Each |
Estimated Shipment: 06-05-2025
Log in to see stock. |
||||||
17533085
![]() |
uvex
9145265 |
Anti-fog, Scratch-resistant |
7.55 €
Each |
In Stock
Log in to see stock. |
||||||
Description
- Reduced to the max - pure, ultralight safety spectacle with innovative side arm concept weighing only 19 grams
- Frameless lens design for a wide field of vision
- X-tended sideshield offers additional protection from hazards
- Flexible side arms deliver the perfect secure, pressure-free fit
- Nose bridge designed for all-day comfort
- Robust polycarbonate lens
- UV400 – 100 % protection against dangerous UVA and UVB radiation up to 400 nm
- Metal-free
- Frame transparent / W 166 FT CE
- Lens PC clear, UV400 / 2C-1,2 W1 FT CE
Specifications
Clear | |
Transparent | |
Reusable | |
Polycarbonate | |
Standard |
Ergonomic Safety Glasses | |
Polycarbonate | |
Polycarbonate | |
UV400 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
uvex Pure-Fit Safety Glasses