missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Block Technology BLOCK™ Pure UV Air and Surface Disinfector
The BLOCK™ Pure UV Disinfector ensures biological decontamination of spaces and is rooted in a combination of HEPA filtration and efficient UV-C lamps. The device operates in two working modes: day Mode - air disinfection in a room, and night Mode - surfaces desinfection in a room.
Brand: Block Technology PUV
This item is not returnable.
View return policy
Description
Intended for
- Hospitals
- Waiting rooms, Consulting rooms
- Locations with heavy human traffic
- Company canteens
- Changing rooms
- Offices, conference rooms
- Retirement homes
- Schools and kindergartens
Advantages
- Captures all impurities from the air with a 99.99% efficiency rate
- Infinitely better compared to simple UV lamps
- Long HEPA filter life
- Long operation life
- Minimum maintenance requirements
- Cleans surfaces for you at night
- Mobile device - ability to move among rooms
- Easy operation
Day Mode
Air disinfection in a room in the presence of personnel.
- The disinfector circulates the air in a room.
- The disinfector is shut down and the air is sucked from the room along the ultraviolet (UV-C) fluorescent tubes.
- Microorganisms inside the disinfector are exposed to strong UV-C radiation for a period of 2 seconds, killing the majority of them.
- The air is filtered with a high-capacity HEPA filter at the disinfector outlet capturing all particles from the air with a 99.99 % efficiency level (i.e. dead and surviving microorganisms are removed from the atmosphere).
- This combination of UV radiation and HEPA filtration achieves near perfect removal of microorganisms from the air.
Night Mode
Surfaces desinfection in a room in the absence of personnel.
- The disinfector is open and the fan is switched off.
- The disinfector is placed near the surfaces that are most exposed and disinfects them with UV-C radiation within the pre-programmed time period.
- TA safety motion sensor completely shuts down the apparatus if anyone enters the radiated space.
- TUV-C radiation is harmful to all living organisms, therefore, rooms must be vacated when Night Mode is in operation. Certain materials are not resistant to UV radiation over the long term.
Specifications
Powder Coated Steel Sheet | |
0.9999 | |
Disinfector |
722 x 1746 x 722 mm | |
Air HEPA Filter, Air Pre-Filter, UV-C Radiation |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction