missing translation for 'onlineSavingsMsg'
Learn More

Labcon™ PurePlus™ 0.1 mL, 96-well PCR Plates with Half Skirt for ABI™ Fast Thermocyclers

These 96-well PCR plates with half skirt are compatible with ABI™ Fast Thermocyclers.

Brand:  Labcon™ 3961-520-000-9

Product Code. 17587946

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • ATP, RNase/DNase and Endotoxin-free (non-pyrogenic)
Specifications

Specifications

Not Autoclavable
Polypropylene
96
100/Cs.
127.8 mm
16 mm
ABI™ Fast Thermocycler
0.1 mL
Non-sterile
10/Pk.
85.5 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers