missing translation for 'onlineSavingsMsg'
Learn More

Labcon™ PurePlus™ 0.2 mL, 8-well PCR Tube Strips with Attached Cap Strips

These 8-well PCR tube strips come bagged with attached cap strips.

Brand:  Labcon™ 3929-550-000-9

Product Code. 17547946

  • 154.00 € / Pack of 125

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Attached clear, flat cap strips
  • ATP, RNase/DNase and Endotoxin-free (non-pyrogenic)
Specifications

Specifications

Clear
125 Tubes/Pk., 10 Pk./Cs.
Polypropylene
69.4 mm
Attached Caps
0.2 mL
Non-sterile
6.5 mm
Not Autoclavable
Bagged
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers