missing translation for 'onlineSavingsMsg'
Learn More

Labcon™ PurePlus™ 1.2 mL Sample Library Tubes

Sample library tubes come in strips of 8, 12, or as individual tubes.

Brand:  Labcon™ 3911-545-000-9

 View more versions of this product

Product Code. 17547936

  • 84.00 € / Pack of 960
Estimated Shipment: 01-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Available in 96-place racks or resealable bags
  • ATP, RNase/DNase, Endotoxin-free (non-pyrogenic)
Specifications

Specifications

Autoclavable
Polypropylene
960/Pk.
12 mL
Clear
Strips of 8, Rack
960 Tubes/Pk., 10 Pk./Cs.
Sterile
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote