missing translation for 'onlineSavingsMsg'
Learn More

Labcon™ PurePlus™ 12-well PCR Tube Strip Caps

These 12-well PCR tube strip caps come conveniently packaged in bags.

Brand:  Labcon™ 3952-540-000-9

Product Code. 17577946

  • 45.90 € / Pack of 80
Estimated Shipment: 19-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • ATP, RNase/DNase and Endotoxin-free (non-pyrogenic)
  • Length (Metric): 114.2 mm
  • Width (Metric): 6.1 mm
Specifications

Specifications

Not Autoclavable
Bagged
80/Pk.
80 Caps/Pk., 10 Pk./Cs.
0.2 mL
Polypropylene
2.5 mm
800/Cs.
Non-sterile
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers