missing translation for 'onlineSavingsMsg'
Learn More

Labcon™ PurePlus™ 25 μL, 384-well PCR Plate with Full Skirt for Popular Thermocyclers

This 384-well PCR plate has a full skirt and is compatible with popular thermocyclers.

Brand:  Labcon™ 3983-520-000-9

Product Code. 17547956

  • 102.00 € / Pack of 10
Estimated Shipment: 19-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • ATP, RNase/DNase and Endotoxin-free (non-pyrogenic)
  • Freezable
Specifications

Specifications

16 x 24
Clear
25 μL
Non-sterile
10/Pk.
127.6 mm
9.6 mm
Not Autoclavable
Polypropylene
384
100/Cs.
10 Plates/Pk., 10 Pk./Cs.
85.7 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers