missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Labcon™ PurePlus™ Cap Strips for 1.2 mL Sample Library Tubes
Complete your 1.2 mL sample library tubes with these cap strips.
33.90 € - 37.75 €
Product Code | Brand | Format | Sterility | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Format | Sterility | Price | Quantity & Availability | ||||
17587936
|
Labcon™
3913-855-000-9 |
Strips of 8 | Sterile |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
17577936
|
Labcon™
3913-850-000-9 |
Strips of 8 | Non-sterile |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
17507946
|
Labcon™
3914-845-000-9 |
Strips of 12 | Sterile |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
17597936
|
Labcon™
3914-840-000-9 |
Strips of 12 | Non-sterile |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- For use with PurePlus™ 1.2 mL Sample Library Tubes
Specifications
PurePlus™ 1.2 mL Sample Library Tubes |