Learn More
Merck PureProteome™ Streptavidin Magnetic Bead System
Suitable for purifying biomolecules, immunoprecipitations, purifying nucleic acids, isolating cells and organelles.
Brand:  Merck LSKMAGT10
Product Code. 12581219
  • 2285.00 € / 10mL
Estimated Shipment: 28-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Bead system helps to purify biotinylated molecules
Recommended Storage
2° to 8°C
TRUSTED_SUSTAINABILITY
Specifications
Specifications
10mL
Magnetic Beads