missing translation for 'onlineSavingsMsg'
Learn More

HALYARD™ PUREZERO™ HG3 Light Blue Sterile Nitrile Gloves
BIOPHARMAGREENER_CHOICE

High-quality gloves designed to the exact standards to meet the requirements of a contamination controlled environment.

437.00 €

Specifications

Certifications/Compliance ISO 3, ISO 4, ISO 5, ISO 6, ISO 7
Certifications/Compliance (Europe) GMP Grade A/B/C/D
Chemotherapeutic Rated ASTM D6978
Chlorinated Yes
Cleanroom Class Class 10 (ISO 4)
View More Specs

Products 8
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
17304350
BIOPHARMAGREENER_CHOICE
View Documents
HALYARD™
CLN923260
6
437.00 €
300 pairs
Estimated Shipment: 15-01-2025
to see stock.
Add to basket
 
17314350
BIOPHARMAGREENER_CHOICE
View Documents
HALYARD™
CLN923265
6.5
437.00 €
300 pairs
In Stock
to see stock.
Add to basket
 
17324350
BIOPHARMAGREENER_CHOICE
View Documents
HALYARD™
CLN923270
7
437.00 €
300 pairs
In Stock
to see stock.
Add to basket
 
17334350
BIOPHARMAGREENER_CHOICE
View Documents
HALYARD™
CLN923275
7.5
437.00 €
300 pairs
In Stock
to see stock.
Add to basket
 
17344350
BIOPHARMAGREENER_CHOICE
View Documents
HALYARD™
CLN923280
8
437.00 €
300 pairs
In Stock
to see stock.
Add to basket
 
17354350
BIOPHARMAGREENER_CHOICE
View Documents
HALYARD™
CLN923285
8.5
437.00 €
300 pairs
In Stock
to see stock.
Add to basket
 
17364350
BIOPHARMAGREENER_CHOICE
View Documents
HALYARD™
CLN923290
9
437.00 €
300 pairs
In Stock
to see stock.
Add to basket
 
17394340
BIOPHARMAGREENER_CHOICE
View Documents
HALYARD™
CLN923210
10
437.00 €
300 pairs
In Stock
to see stock.
Add to basket
 
Description

Description

A cleanroom glove with guaranteed low particulate counts and endotoxin levels due to repeated washes in deionized water and chlorination.

  • Nickel-free
  • Powder-free
  • Silicone-free
  • Latex-free and no Phtalates
  • High cleanliness
  • Low endotoxin levels
  • Formulated with ZERO presence of these hazardous traditional accelerators: Sulfur, Thiurams, Thiazoles, Guanidine, Carbamates(ZDEC or ZDBC)
  • Backed up by a vertically integrated supply chain
Specifications

Specifications

ISO 3, ISO 4, ISO 5, ISO 6, ISO 7
ASTM D6978
Class 10 (ISO 4)
1
Single-use
4 mil
Specific
Nitrile
0.08 mm
Controlled Environment Gloves
4 mil
ISO 9001:2015, EN13485:2016
30.5 cm
GMP Grade A/B/C/D
Yes
Yes
Light Blue
Partially Textured
0.10 mm
Yes
3.1 mil
Powder Free
Sterile
0.10 mm
12 in.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.