missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HALYARD™ PUREZERO™ HG3 White Sterile Nitrile Gloves
High-quality gloves designed to the exact standards to meet the requirements of a contamination controlled environment.
398.00 €
Specifications
Certifications/Compliance | ISO 3, ISO 4, ISO 5, ISO 6, ISO 7 |
---|---|
Certifications/Compliance (Europe) | GMP Grade A/B/C/D |
Chemotherapeutic Rated | ASTM D6978 |
Chlorinated | Yes |
Cleanroom Class | Class 10 (ISO 4) |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
17374330
|
HALYARD™
CLN323260 |
6 |
398.00 €
200 pairs |
Estimated Shipment: 16-12-2024
Log in to see stock. |
|||||
17384330
|
HALYARD™
CLN323265 |
6.5 |
398.00 €
200 pairs |
In Stock
Log in to see stock. |
|||||
17394330
|
HALYARD™
CLN323270 |
7 |
398.00 €
200 pairs |
In Stock
Log in to see stock. |
|||||
17304340
|
HALYARD™
CLN323275 |
7.5 |
398.00 €
200 pairs |
In Stock
Log in to see stock. |
|||||
17314340
|
HALYARD™
CLN323280 |
8 |
398.00 €
200 pairs |
In Stock
Log in to see stock. |
|||||
17324340
|
HALYARD™
CLN323285 |
8.5 |
398.00 €
200 pairs |
In Stock
Log in to see stock. |
|||||
17334340
|
HALYARD™
CLN323290 |
9 |
398.00 €
200 pairs |
Estimated Shipment: 17-12-2024
Log in to see stock. |
|||||
17364330
|
HALYARD™
CLN323210 |
10 |
398.00 €
200 pairs |
In Stock
Log in to see stock. |
|||||
Description
A cleanroom glove with guaranteed low particulate counts and endotoxin levels due to repeated washes in deionized water and chlorination.
- Nickel-free
- Powder-free
- Silicone-free
- Latex-free and no Phtalates
- High cleanliness
- Low endotoxin levels
- Formulated with ZERO presence of these hazardous traditional accelerators: Sulfur, Thiurams, Thiazoles, Guanidine, Carbamates(ZDEC or ZDBC)
- Backed up by a vertically integrated supply chain
Specifications
ISO 3, ISO 4, ISO 5, ISO 6, ISO 7 | |
ASTM D6978 | |
Class 10 (ISO 4) | |
1 | |
Single-use | |
6 mil | |
Specific | |
Nitrile | |
0.13 mm | |
Controlled Environment Gloves | |
6 mil | |
ISO 9001:2015, EN13485:2016 | |
30.5 cm |
GMP Grade A/B/C/D | |
Yes | |
Yes | |
White | |
Partially Textured | |
0.16 mm | |
Yes | |
5 mil | |
Powder Free | |
Sterile | |
0.16 mm | |
12 in. |