Learn More
Fisherbrand™ Prefiltration Cartridge
Treats, softens, purifies, and conditions tap water.. Fisher Scientific™ Prefiltration Cartridges are used with the Accu500 Ultrapure Water System.
121.00 € - 191.00 €
Specifications
For Use With (Equipment) Accu500 Ultrapure Water System
Height (Metric) 254 mm
View More Specs
Products 2
Product Code Brand Type Price Quantity & Availability  
Product Code Brand Type Price Quantity & Availability  
15489639
View Documents
Fisherbrand™
LABAC1012
Activated Carbon Cartridge
121.00 €
Pack of 12
Estimated Shipment: 28-05-2025
to see stock.
Add to basket
15499639
View Documents
Fisherbrand™
LABAT1012
Anti-scaling Softening Cartridge
191.00 €
Pack of 12
Estimated Shipment: 12-06-2025
to see stock.
Add to basket
Description
Description
  • Anti-Scaling Softener Cartridge treats and conditions hard water
  • Activated Carbon Cartridge removes particles, organics and chlorine
Specifications
Specifications
Accu500 Ultrapure Water System
254 mm