missing translation for 'onlineSavingsMsg'
Learn More

Euro Protection™ Purofort™ Safety Boots

Euro Protection™ Purofort™ Safety Boots provide flexible and lightweight protection and cold insulation and feature a steel toe cap for protection.

97.90 € - 104.00 €

Specifications

Color Green
For Use With (Application) Agriculture/Farming
Liner Material Polyamide
Sole Features Polyurethane insole, steel, puncture-resistant sole, Purofort™ outsole.
Sole Material Purofort™
View More Specs

Products 2
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
15869226
View Documents
Euro Protection™
9PUSG39
39 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15879226
View Documents
Euro Protection™
9PUSG40
40 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Flexible and lightweight for use all day long
  • Cold insulation
  • Tear and wear resistant
  • Purofort™ upper
  • Polyamide lining
  • Polyurethane insole
  • Steel toe cap
  • Steel, puncture-resistant sole
  • Purofort™ outsole
  • Suitable for use in farming industry
Specifications

Specifications

Green
Polyamide
Purofort™
900 g per boot in size 9.5
Safety Boot
Agriculture/Farming
Polyurethane insole, steel, puncture-resistant sole, Purofort™ outsole.
Steel
EN ISO 20345 : 2011
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.