Learn More
Puromycin Dihydrochloride, Fisher BioReagents™
Inhibits protein synthesis
Brand:  Fisher BioReagents BP2956-100
Product Code. 10054207
  • 152.00 € / 100mg
Explore more special offers
Description
Description
Inhibits protein synthesis
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Puromycin Dihydrochloride
Solid
168°C
> 98 %
MFCD00150080
Clear Glass
Cl.Cl.COC1=CC=C(C[C@H](N)C(=O)N[C@@H]2[C@@H](CO)O[C@H]([C@@H]2O)N2C=NC3=C(N=CN=C23)N(C)C)C=C1
544.43
544.44
<12.0
100 mg
Puromycin Dihydrochloride
58-58-2
Streptomyces alboniger
C22H31Cl2N7O5
15, 8054
MKSVFGKWZLUTTO-MLYJQVMKNA-N
(2S)-2-amino-N-[(2S,3S,4S,5R)-5-[6-(dimethylamino)purin-9-yl]-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl]-3-(4-methoxyphenyl)propanamide;dihydrochloride
131632508
98% min.
Beige
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Puromycin dihydrochloride hydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.