missing translation for 'onlineSavingsMsg'
Learn More

PURUS™ Frames for Biohybrid™ Contamination Control Mats

Designed to be used with the PURUS™ Biohybrid™ mat to create a system that is easy to use in any critical area where direct adhesion of mats to a flooring surface is not allowed.

Brand:  PURUS™ PF 2638 01 W 00

Product Code. 15362002

  • 164.00 € / Each
Estimated Shipment: 08-07-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Easily moved for cleaning
  • Non-skid backing ensures frame stays in place
  • Fits all PURUS™ Biohybrid™ mats
  • Strong and Durable
  • Non-Slip made from Polymer Coated polyester textile
  • Naturally antimicrobial, mold and mildew resistant
Specifications

Specifications

660 x 965 mm
610 x 914 mm PURUS™ Biohybrid™ Mats
White
For use in Pharmaceutical, Biotech, Semiconductor, Cleanrooms, and Industrial Applications
3.175 mm
Polystyrene
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
PURUS™ Frames for Biohybrid™ Contamination Control Mats > 660 x 965 mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.