Learn More
Fisherbrand™ Push-in Cap Glass Specimen Vials
Store and display specimens securely and easily. Fisherbrand™ Push-in Cap Glass Specimen Vials are available in a variety of sizes. Fisherbrand™ screw-in polyethylene caps are sold separately.
Brand:  Fisherbrand™ 15314779
Product Code. 15314779
  • 174.00 € / Pack of 882
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Manufactured from clear Type 1B neutral glass
  • Versatile capacity range from a variety of sizes
  • 1 dram equates to approximately 3.55mL (or 0.125 fl. oz.)
  • Suitable for media, diagnostic, storage, display, and sample collection applications
  • Fisherbrand™ Polyethylene Push-in Caps for Glass Specimen Vials sold separately
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Clear
Neutral Glass (Type 1B)
Push-in Cap
8 mm
11 mm
1.75 mL
36 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products