missing translation for 'onlineSavingsMsg'
Learn More
Ansell Edmont™ AlphaTec™ 65-100 PVC Aprond Product Code.: 15643146
Add to basket

Ansell Edmont™ AlphaTec™ 65-100 PVC Aprond

Product Code. 15643146 Shop All Ansell Products
Green
Click to view available options
Green
This item is not returnable. View return policy

Product Code. 15643146

Brand: Ansell Edmont™ 56100

Add to basket
Add to basket
This item is not returnable. View return policy

Offer excellent protection where splashes from acids and caustic bases are a concern.

  • PVC construction is flexible yet offers resistance to abrasion and puncture
  • Sealed grommets with adjustable tiesallow for size adjustment without tear-out

Order Info

dimColor

TRUSTED_SUSTAINABILITY

Specifications

Type Apron
Material PVC
Color Green
Resistance Chemicals, fats, oils, and grease
Thickness 0.508 mm
Includes An extra-long neck and waist ties.
For Use With (Application) Food handling and processing industrial maintenance and chemical processing.Â
Dimensions (L x W) 35 x 45 in.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ansell Edmont™ AlphaTec™ 65-100 PVC Aprond >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.