Learn More
Cole-Parmer™ Digi-Sense™ Type-T PVC-Insulated Probes with Epoxy-Coated Tip
Use these probes for applications requiring an isolated junction. Digi-Sense™ Type-T PVC-Insulated Probes with Epoxy-Coated Tip are ideal for short term immersion.
Brand:  Cole-Parmer™ 08466-06
Product Code. 11731349
  • 33.65 € / Each
Estimated Shipment: 10-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Each probe comes with a color coded ANSI mini connector: type J-black, type K-yellow, type T-blue and type E-purple. Response Time: 2 seconds or greater.
  • Exposed-junction probes are not recommended for use with aqueous solutions
  • Do not subject type J exposed-junction probes to moisture as they will rust
Specifications
Specifications
ANSI
Thermocouple Probe
20.3 cm
-200°C to +105°C
Used for Short Term Immersion