Learn More
Cole-Parmer™ PVC Stopcocks with Flow Indication
Eliminate guess work with these stopcocks with flow indication. Cole Parmer™ PVC Stopcocks with Flow Indication feature PVC bodies, EPDM seals and seats. Flow indicator on handle.
Brand:  Cole-Parmer™ 06225-54
 View more versions of this product
Product Code. 11809442
  • 47.20 € / Each
Estimated Shipment: 28-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Maximum pressure is 150psi at the maximum temperature of 49°C
  • Maximum vacuum is 29.9'' Hg
Specifications
Specifications
PVC
0.5 to 5/8 in.
1 Each
150 psi
1 Each
NPT(M) x Hose