missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ PVC Stopcocks with Flow Indication

Eliminate guess work with these stopcocks with flow indication. Cole Parmer™ PVC Stopcocks with Flow Indication feature PVC bodies, EPDM seals and seats. Flow indicator on handle.

Brand:  Cole-Parmer™ 06225-56

 View more versions of this product

Product Code. 11809452

  • 46.60 € / Each
Estimated Shipment: 28-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Maximum pressure is 150psi at the maximum temperature of 49°C
  • Maximum vacuum is 29.9'' Hg
Specifications

Specifications

PVC
0.25 to 0.5 in.
1 Each
150 psi
1 Each
Hose x Hose
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.