Learn More
Clark Scientific™ PVC Large White Spill Trays
Manufactured from rigid PVC, these large white plastic spill trays are for use outside radiation areas.
Brand:  Clark Scientific™ WPVC large
 View more versions of this product
Product Code. 12988837
  • 246.00 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
    • Manufactured from rigid PVC
    • Available in yellow for use in radiation areas
    • Available in white for use outside radiation areas
    • Colourless PVC liners are available
Specifications
Specifications
White
Spill Tray Bench
1 Ea.
450 mm
PVC
Outside radiation areas
700 mm