missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Masterflex™ PVDF Straight Barbed Union
Specifications
Product Type | Straight Barbed Union |
---|---|
Material | PVDF |
Min. Operating Temperature (Metric) | -62°C |
Packaging Quantity | 10 Pack |
Max. Operating Temperature (Metric) | 129°C |
Specifications
Straight Barbed Union | |
-62°C | |
129°C |
PVDF | |
10 Pack |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title