missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PVR (CD155) Mouse anti-Human, BUV737, Clone: SKII.4, BD Biosciences™
Mouse Monoclonal Antibody
Brand: BD Biosciences 748586
This item is not returnable.
View return policy
Description
PVR (CD155) Monoclonal antibody specifically detects PVR (CD155) in Human samples. It is validated for Flow CytometrySpecifications
PVR (CD155) | |
Monoclonal | |
Brilliant Ultraviolet 737 | |
poliovirus receptor; HVED; nectin-like protein 5; NECL5; Necl-5; PVR; PVS; TAGE4 | |
SK-N-AS neuroblastoma cell line | |
RUO | |
Human | |
IgG1 κ |
Flow Cytometry | |
SKII.4 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV737 under optimal conditions that minimize unconjugated dye and antibody. |