missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ PX 12

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Thioredoxin-1 (Trx-1) inhibitor; irreversibly inactivates Trx-1 as a substrate for reduction by thioredoxin reductase (TR); competitively inhibits the reduction of Trx-1 by TR (Ki = 30.8 μM). Attenuates expression of HIF-1α, VEGF and iNOS (IC50 values are 7.2, 10.4 and 18.1 μM respectively). Exhibits antitumor activity; directly inhibits tubulin in vitro and decreases tumor microvessel density in vivo.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 188.31 |
| Purity | >99% |
| Molecular Weight (g/mol) | 188.31 |
| Chemical Name or Material | PX 12 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur