missing translation for 'onlineSavingsMsg'
Learn More
Ansell PX130 Lightweight Polyurethane-Coated Nylon Glove Liners Product Code.: 16895330
Add to basket

Ansell PX130 Lightweight Polyurethane-Coated Nylon Glove Liners

Product Code. 16895330 Shop All Ansell Products
6
Click to view available options
Size:
6
7
8
9
10
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Product Code. 16895330

Brand: Ansell C382060

Add to basket
Add to basket
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Polyurethane coating on the palm provides abrasion resistance, with an elasticated knit wrist to ensure a snug fit.

  • Polyurethane coating on the palm provides abrasion resistance
  • Elasticated knit wrist ensures a secure fit
  • Ventilated back is breathable to minimize perspiration
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 3
Certifications/Compliance (Europe) EN 420:2003 + A1:2009; Category II
Coated Area Palm
Coating Material Polyurethane
Color Gray
Cut Resistance Rating ANSI A1
Hand Specific
Material Nylon
Size 6
For Use With (Application) Automotive Assembly, Machinery, and Equipment Operation
Glove Cuff Style Knit
Series PX130
Yarn Gauge 15
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ansell PX130 Lightweight Polyurethane-Coated Nylon Glove Liners >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.