missing translation for 'onlineSavingsMsg'
Learn More

PYBOP(R), 99%, Thermo Scientific Chemicals

122.61 € - 388.94 €

Chemical Identifiers

CAS 128625-52-5
Molecular Formula C18H28F6N6OP2
Molecular Weight (g/mol) 520.39
MDL Number MFCD00077411
InChI Key VIAFLMPQBHAMLI-UHFFFAOYSA-N
Synonym pybop, 1h-benzo d 1,2,3 triazol-1-yl oxy tri pyrrolidin-1-yl phosphonium hexafluorophosphate v, benzotriazol-1-yloxy tripyrrolidinophosphonium hexafluorophosphate, benzotriazole-1-yl-oxytripyrrolidinophosphonium hexafluorophosphate, benzotriazol-1-yl-oxytripyrrolidinophosphonium hexafluorophosphate, 1h-benzotriazol-1-yloxytripyrrolidinophosphonium hexafluorophosphate, benzotriazol-1-yloxytris pyrrolidino phosphonium hexafluorophosphate, benzotriazole-1-yl-oxy-tris-pyrrolidino-phosphonium hexafluorophosphate, benzotriazol-1-yl-oxy-tripyrrolidinophosphonium hexafluorophosphate, benzotriazol-1-yl-oxytripyrrolidinphosphonium hexafluorophosphate
PubChem CID 2724699
IUPAC Name benzotriazol-1-yloxy(tripyrrolidin-1-yl)phosphanium;hexafluorophosphate
SMILES C1CCN(C1)[P+](N2CCCC2)(N3CCCC3)ON4C5=CC=CC=C5N=N4.F[P-](F)(F)(F)(F)F
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10581311
SDS View Documents
Thermo Scientific Acros
270720050
5 g
122.61 €
5g
Please to purchase this item. Need a web account? Register with us today!
10750951
SDS View Documents
Thermo Scientific Acros
270720250
25 g
388.94 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
Specifications

Chemical Identifiers

128625-52-5
520.39
VIAFLMPQBHAMLI-UHFFFAOYSA-N
2724699
C1CCN(C1)[P+](N2CCCC2)(N3CCCC3)ON4C5=CC=CC=C5N=N4.F[P-](F)(F)(F)(F)F
C18H28F6N6OP2
MFCD00077411
pybop, 1h-benzo d 1,2,3 triazol-1-yl oxy tri pyrrolidin-1-yl phosphonium hexafluorophosphate v, benzotriazol-1-yloxy tripyrrolidinophosphonium hexafluorophosphate, benzotriazole-1-yl-oxytripyrrolidinophosphonium hexafluorophosphate, benzotriazol-1-yl-oxytripyrrolidinophosphonium hexafluorophosphate, 1h-benzotriazol-1-yloxytripyrrolidinophosphonium hexafluorophosphate, benzotriazol-1-yloxytris pyrrolidino phosphonium hexafluorophosphate, benzotriazole-1-yl-oxy-tris-pyrrolidino-phosphonium hexafluorophosphate, benzotriazol-1-yl-oxy-tripyrrolidinophosphonium hexafluorophosphate, benzotriazol-1-yl-oxytripyrrolidinphosphonium hexafluorophosphate
benzotriazol-1-yloxy(tripyrrolidin-1-yl)phosphanium;hexafluorophosphate

Specifications

128625-52-5
White
98.5% min. (HPLC)
C18H28F6N6OP2
pybop, 1h-benzo d 1,2,3 triazol-1-yl oxy tri pyrrolidin-1-yl phosphonium hexafluorophosphate v, benzotriazol-1-yloxy tripyrrolidinophosphonium hexafluorophosphate, benzotriazole-1-yl-oxytripyrrolidinophosphonium hexafluorophosphate, benzotriazol-1-yl-oxytripyrrolidinophosphonium hexafluorophosphate, 1h-benzotriazol-1-yloxytripyrrolidinophosphonium hexafluorophosphate, benzotriazol-1-yloxytris pyrrolidino phosphonium hexafluorophosphate, benzotriazole-1-yl-oxy-tris-pyrrolidino-phosphonium hexafluorophosphate, benzotriazol-1-yl-oxy-tripyrrolidinophosphonium hexafluorophosphate, benzotriazol-1-yl-oxytripyrrolidinphosphonium hexafluorophosphate
VIAFLMPQBHAMLI-UHFFFAOYSA-N
benzotriazol-1-yloxy(tripyrrolidin-1-yl)phosphanium;hexafluorophosphate
2724699
99%
PYBOP*, 99%
145°C to 160°C
Authentic
Glass bottle
MFCD00077411
Solubility in water: .. Other solubilities: soluble in acetonitrile,dmf,dcm,thf and,n-methylpyrrolidine
C1CCN(C1)[P+](N2CCCC2)(N3CCCC3)ON4C5=CC=CC=C5N=N4.F[P-](F)(F)(F)(F)F
520.39
520.39
Fine Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

Product Identifier
  • PYBOP

Signal Word
  • Warning

Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.