missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lenz Laborglasintrumente Pycnometers acc. To Gay Lussac Unadjusted
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
33.00 € - 38.00 €
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10797701
|
Lenz Laborglasintrumente
05630010 |
5 mL |
33.00 €
Each |
Estimated Shipment: 21-02-2025
Log in to see stock. |
|||||
10315691
|
Lenz Laborglasintrumente
05630013 |
10 mL |
38.00 €
Each |
Estimated Shipment: 21-02-2025
Log in to see stock. |
|||||
Description
- Gay Lussac pycnometers acc. to DIN ISO 3507
- Unadjusted, stopper NS 10/19
- With capillary, stopper top ground and polished
- Imprint of nominal capacity at the bottom
- Material: borosilicate glass 3.3
Specifications
Borosilicate Glass (3.3) |