Learn More
Karl Hecht™ Pycnometers with Ground-In Thermometer
For the measurement of liquid density and temperature in the laboratory.
Brand:  Karl Hecht™ 42572032
Product Code. 10504432
  • 224.00 € / Each
Explore more special offers
Description
Description
  • With ground-in thermometer
  • Precisely adjusted
  • Side capillary with ground-on cap joint size 7.5
  • Flask of Erlenmeyer form
  • Thermometer ground joint size 10/19
  • Thermometer with blue filling, +10°C + 35°C
Specifications
Specifications
Pycnometer
Measuring liquid density, temperature
With ground-in thermometer
50 mL
Blue thermometer filling