Learn More
Tocris Bioscience™ Pyr 10
Selective TRPC3 inhibitor
Brand:  Tocris Bioscience™ 6941/50
539.08 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18768946
  • 682.00 € / 50mg
Explore more special offers
Description
Description
Pyr 10 is a TRPC3 (transient receptor potential canonical 3) channel inhibitor (IC50 = 0.72 μM for TRPC3 mediated calcium entry in vitro), which displays approximately 18-fold selectivity for TRPC3-mediated receptor operated calcium entry (ROCE) over STIM1/Orai1-mediated store operated calcium entry (SOCE). Pyr 10 inhibits NFATc3 activation and inhibits proliferation of rat ventricular cardiac fibroblasts.
Specifications
Specifications
N-[4-[3,5-bis(Trifluoromethyl)-1 H-pyrazol-1-yl]phenyl]-4-methylbenzenesulfonamide
50 mg
C18H13F6N3O2S
1315323-00-2
TRPC Channel Blockers
0.98