missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Pyr3
GREENER_CHOICE

Product Code. 18043782
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18043782

Brand: Tocris Bioscience™ 3751/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective TRPC3 blocker

Selective antagonist of the canonical transient receptor potential channel 3 (TRPC3). Inhibits TRPC3-mediated Ca2+ influx (IC50 = 0.7 μM) and suppresses activation of nuclear factor of activated T cells (NFAT). Inhibits hypertrophic responses in cardiomyocytes.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1160514-60-2
Molecular Formula C16H11Cl3F3N3O3

Specifications

Quantity 50 mg
Formula Weight 456.63
Purity >98%
Molecular Weight (g/mol) 456.627
Chemical Name or Material Pyr3
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.