missing translation for 'onlineSavingsMsg'
Learn More
Pyrazolo[1,5-a]pyridine-2-carboxylic acid, Tech., Thermo Scientific™ Product Code.: 10544493

Pyrazolo[1,5-a]pyridine-2-carboxylic acid, Tech., Thermo Scientific™

Product Code. 10544493
1 g, Amber Glass Bottle
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10544493

Brand: Thermo Scientific Maybridge CC39401DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 63237-88-7
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.15
MDL Number MFCD06739057
InChI Key AYGNAOBAPQNTIL-UHFFFAOYSA-N
Synonym pyrazolo 1,5-a pyridine-2-carboxylic acid, 2-carboxypyrazolo 1,5-a pyridine, h-pyrazolo 1,5-a pyridine-2-carboxylic acid, pubchem15887, 2-pyrazolo 1,5-a pyridinecarboxylic acid, 8-hydropyrazolo 1,5-a pyridine-2-carboxylic acid, pyrazolo 1 pound not5-a pyridine-2-carboxylicacidShow MoreShow Less
PubChem CID 7127820
IUPAC Name pyrazolo[1,5-a]pyridine-2-carboxylic acid
SMILES OC(=O)C1=NN2C=CC=CC2=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 162.15
Percent Purity 97%
Grade Technical
Chemical Name or Material Pyrazolo[1, 5-a]pyridine-2-carboxylic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Pyrazolo[1,5-a]pyridine-2-carboxylic acid, Tech., Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.