missing translation for 'onlineSavingsMsg'
Learn More
Pyrazolo[1,5-a]pyridine, 97%, Thermo Scientific Chemicals Product Code.: 15497527

Pyrazolo[1,5-a]pyridine, 97%, Thermo Scientific Chemicals

Product Code. 15497527
1 g
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15497527

Brand: Thermo Scientific Alfa Aesar H35502.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 274-56-6
Molecular Formula C7H6N2
Molecular Weight (g/mol) 118.14
MDL Number MFCD08752622
InChI Key DVUBDHRTVYLIPA-UHFFFAOYSA-N
Synonym pyrazolo 1,5-a pyridine, 3-azaindoline, pyrazolo 2,3-a pyridine, pyrazolo 1,5-a-pyridine, 3-azaindolizine, pyrazolo 1,5-apyridine, h-pyrazolo 1,5-a pyridineShow MoreShow Less
PubChem CID 67507
IUPAC Name pyrazolo[1,5-a]pyridine
SMILES C1=NN2C=CC=CC2=C1

Specifications

Color Brown or Colorless to Red
Boiling Point 37°C to 39°C (0.2 mmHg)
Refractive Index 1.6075
Quantity 1 g
Formula Weight 118.14
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Pyrazolo[1,5-a]pyridine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10175516 Certificate TypeCertificate of Analysis Date16/04/2024 Catalog NumberH35502.03,H35502.MD
Lot Number10175516 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH35502.03,H35502.MD

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Pyrazolo[1,5-a]pyridine, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.