Learn More
Pyrex™ Borosilicate Glass Erlenmeyer Flask with Quickfit Ground Glass Socket
Grow cultures and store other laboratory reagents in Glass Erlenmeyer flasks. The Pyrex™ borosilicate glass Erlenmeyer flask with QuickFit™ glass socket is sturdy and comes in a variety of sizes.
Brand:  Pyrex 1132/06
 View more versions of this product
Product Code. 15507670
  • 544.00 € / Pack of 20
Estimated Shipment: 22-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Grow cultures and store other laboratory reagents in Glass Erlenmeyer flasks. The Pyrex™ borosilicate glass Erlenmeyer flask with QuickFit™ glass socket is sturdy and comes in a variety of sizes.
  • Complies to ISO 4797
  • Conical flasks with highest quality Quickfit™ ground glass socket
  • Borosilicate glass for corrosion free performance and high resistance to thermal shock
  • Erlenmeyer flasks with high quality Quickfit™ ground glass socket
  • Manufactured from Pyrex™ borosilicate glass for highest resistance to thermal shock and chemical corrosion
  • White enamel graduations and marking spot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DIN 12387
Borosilicate Glass
110 mm
29/32
Yes
100 mL
Yes
Conical
Yes
64 mm