missing translation for 'onlineSavingsMsg'
Learn More

Pyrex™ Borosilicate Glass Narrow Mouth Reagent Bottles with Glass Stopper

Store reagents, analytical standards and culture media in these glass reagent bottles. Pyrex™ Borosilicate Glass Narrow Mouth Reagent Bottles with Glass Stopper features excellent chemical resistance and can withstand high temperatures.

Brand:  Pyrex 1521/18D

323.95 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 11347864

  • 350.00 € / Each
Estimated Shipment: 05-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Complies with ISO 4796-2
  • Supplied with solid glass stopper
  • Pyrex™ borosilicate glass with ground neck
  • Autoclavable
TRUSTED_SUSTAINABILITY
Specifications

Specifications

10,000 mL
Borosilicate Glass
ISO 4796-2
Narrow
Glass
227 mm
60 mm
1/PK
Yes
414 mm
60/46 mm
Stopper
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers