missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrex™ Borosilicate Glass Narrow Mouth Reagent Bottles with Glass Stopper
Store reagents, analytical standards and culture media in these glass reagent bottles. Pyrex™ Borosilicate Glass Narrow Mouth Reagent Bottles with Glass Stopper features excellent chemical resistance and can withstand high temperatures.
Brand: Pyrex 1521/12D
2873.75 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Complies with ISO 4796-2
- Supplied with solid glass stopper
- Pyrex™ borosilicate glass with ground neck
- Autoclavable
Specifications
2000 mL | |
Borosilicate Glass | |
ISO 4796-2 | |
Narrow | |
Glass | |
138 mm |
29 mm | |
10/PK | |
Yes | |
262 mm | |
29/32 mm | |
Stopper |