Learn More
Pyrex™ Borosilicate Glass Narrow Neck Flat Bottom Boiling Flask
Manufactured from Pyrex™ glass for high resistance to thermal shock. Pyrex™ Borosilicate Glass Narrow Neck Flat Bottom Boiling Flask complies with ISO 1773 standards. Excellent chemical durability for contamination-free performance. Includes white enamel marking spot while flat base prevents spillage.
Brand:  Pyrex 1070/28D
 View more versions of this product
Product Code. 10801601
  • 458.00 € / Pack of 10
Estimated Shipment: 07-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Complies with ISO 1773
  • Flat base for standing on benches and hotplates etc
  • Manufactured from Pyrex™ glass for high resistance to thermal shock
  • Excellent chemical durability for contamination-free performance
  • With white enamel marking spot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ISO 1773
2000 mL
250 mm
Flat
45 mm
Borosilicate Glass
Narrow
166 mm