Learn More
Pyrex™ Borosilicate Glass Narrow Neck Round Bottom Boiling Flask
Resistant to chemical corrosion and thermal shock
12.47 € - 483.00 €
Specifications
Certifications/Compliance | ISO 1773 |
---|---|
Material | Borosilicate Glass |
Bottom Shape | Round |
Product Code | Brand | Capacity (Metric) | Diameter (Metric) Outer | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Diameter (Metric) Outer | Price | Quantity & Availability | ||||
10327899
|
Pyrex
1100/36D |
10 L | 279 mm |
269.00 €
Each |
Estimated Shipment: 16-04-2025
Log in to see stock. |
||||
15157203
|
Pyrex
1100/04M |
100 mL | 65 mm | N/A | |||||
12579095
|
Pyrex
1100/04M |
100 mL | 65 mm |
154.00 €
Pack of 10 |
Estimated Shipment: 07-03-2025
Log in to see stock. |
||||
10587670
|
Pyrex
1100/38D |
20 L | 345 mm |
483.00 €
Each |
Estimated Shipment: 16-04-2025
Log in to see stock. |
||||
10754972
|
Pyrex
1100/28D |
2000 mL | 166 mm |
56.90 €
Each |
In Stock
Log in to see stock. |
||||
10079139
|
Pyrex
1100/32D |
4 L | 207 mm |
132.00 €
Each |
Estimated Shipment: 16-04-2025
Log in to see stock. |
||||
12569095
|
Pyrex
1100/02M |
50 mL | 50 mm |
122.00 €
Pack of 10 |
Estimated Shipment: 24-01-2025
Log in to see stock. |
||||
10089139
|
Pyrex
1100/35D |
6 L | 236 mm |
174.00 €
Each |
Estimated Shipment: 16-04-2025
Log in to see stock. |
||||
12099002
|
Pyrex
1100/12D |
250 mL | 85 mm |
160.00 €
Pack of 10 |
Estimated Shipment: 21-03-2025
Log in to see stock. |
||||
12009012
|
Pyrex
1100/18D |
500 mL | 105 mm |
171.00 €
Pack of 10 |
Estimated Shipment: 07-02-2025
Log in to see stock. |
||||
12019012
|
Pyrex
1100/24D |
1000 mL | 131 mm |
277.00 €
Pack of 10 |
Estimated Shipment: 17-01-2025
Log in to see stock. |
||||
Description
- Complies with ISO 1773
- Manufactured from Pyrex™ borosilicate glass which offers excellent resistance to chemical corrosion and thermal shock
Specifications
ISO 1773 | |
Borosilicate Glass | |
Round |