missing translation for 'onlineSavingsMsg'
Learn More

Pyrex™ Borosilicate Glass Non-vacuum Desiccators

Simple design for drying under atmospheric pressure.

Brand:  Pyrex 1591/02D

 View more versions of this product

Product Code. 12476947

  • 496.00 € / Each
Estimated Shipment: 10-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Robust desiccator base and cover in borosilicate glass
  • Ground glass flanges
  • Plates are positioned on an internal ledge within the base
  • Choice of 2 sizes - to accommodate 150mm or 200mm diameter plates
  • Accessory metal perforated plate available
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Nonvacuum Desiccator
Plate: 200 mm
220 mm
Complete desiccator
120 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.