Learn More
Pyrex™ Borosilicate Glass Plain Neck Kjeldahl Flask
Suitable for the determination of nitrogen
Brand:  Pyrex 1200/10D
641.25 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 15547680
  • 692.00 € / Pack of 20
Estimated Shipment: 04-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Withstands high temperatures
  • Resistant to chemical corrosion
TRUSTED_SUSTAINABILITY
Specifications
Specifications
24 mm
300 mL
300 mm
81 mm
28 mm
Borosilicate Glass
Plain