missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrex™ Borosilicate Glass Reagent Bottles
Click to view available options
Capacity (Metric):
10,000 mL
100 mL
1000 mL
15,000 mL
20,000 mL
2000 mL
25 mL
250 mL
50 mL
500 mL
5000 mL
Description
Store reagents, analytical standards and culture media in these borosilicate glass reagent bottles. PyrexTM Borosilicate Glass Reagent Bottles withstand hot air sterilization up to 180°C, and have excellent chemical resistance.
- Complies with ISO 4796-1
- Manufactured from Pyrex™ borosilicate glass
- Excellent resistance to chemical attack and high temperatures
- Supplied with PBT screw caps that have a PTFE faced sealing disc
- Autoclavable
- White graduations and marking spot
Specifications
Specifications
Capacity (Metric) | 5000 mL |
For Use With (Application) | Ideal for storage of reagents, analytical standards and culture media etc |
Material | Borosilicate Glass |
No. per Pack | 1/PK |
Closure Color | Black |
Certifications/Compliance | ISO 4796-1 |
Closure Included | Yes |
Autoclavable | Autoclavable |
Graduated | Yes |
Height (Metric) | 324 mm |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction