Learn More
Pyrex™ Borosilicate Glass, Wide-Neck, Round Media Bottles
Manufactured from temperature and chemically resistant borosilicate glass and complies with ISO 479. Pyrex™ Borosilicate Glass Wide Neck Round Media Bottles feature sloping shoulders for smooth pouring action. White graduations and marking spot provide easy volume assessment.
61.00 € - 605.00 €
Specifications
Color Clear
Material Borosilicate Glass
Certifications/Compliance ISO 4796
Closure Included No
Autoclavable Autoclavable
View More Specs
Products 11
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
11333504
View accessory products View Documents
Pyrex
1517/01
25 mL
77.55 €
Pack of 10
Estimated Shipment: 31-03-2025
to see stock.
Add to basket
11343504
View accessory products View Documents
Pyrex
1517/02
50 mL
72.80 €
Pack of 10
Estimated Shipment: 14-04-2025
to see stock.
Add to basket
11353504
View accessory products View Documents
Pyrex
1517/04
100 mL
61.00 €
Pack of 10
Estimated Shipment: 24-02-2025
to see stock.
Add to basket
15757927
View Documents
Pyrex
1517/05
150 mL
127.00 €
Pack of 10
Estimated Shipment: 31-03-2025
to see stock.
Add to basket
11363504
View accessory products View Documents
Pyrex
1517/06
250 mL
71.75 €
Pack of 10
Estimated Shipment: 31-03-2025
to see stock.
Add to basket
11373504
View accessory products View Documents
Pyrex
1517/08
500 mL
101.00 €
Pack of 10
Estimated Shipment: 13-02-2025
to see stock.
Add to basket
11383504
View accessory products View Documents
Pyrex
1517/10
1000 mL
150.00 €
Pack of 10
Estimated Shipment: 03-02-2025
to see stock.
Add to basket
11393504
View accessory products View Documents
Pyrex
1517/12
2000 mL
394.00 €
Pack of 10
Estimated Shipment: 18-03-2025
to see stock.
Add to basket
15777927
View Documents
Pyrex
1517/14
3500 mL
605.00 €
Each
Estimated Shipment: 31-03-2025
to see stock.
Add to basket
11303514
View accessory products View Documents
Pyrex
1517/16
5000 mL
184.00 €
Each
Estimated Shipment: 17-02-2025
to see stock.
Add to basket
11313514
View accessory products View Documents
Pyrex
1517/18
10,000 mL
333.00 €
Each
Estimated Shipment: 21-05-2025
to see stock.
Add to basket
Description
Description
  • Bottles supplied without cap or pouring ring
  • Sloping shoulders for smooth pouring action
  • White graduations and marking spot
  • Manufactured from temperature and chemically resistant borosilicate glass
  • Complies with ISO 4796
  • Printed with Trace Code for downloadable batch certificate
Specifications
Specifications
Clear
ISO 4796
Autoclavable
Yes
Round
Borosilicate Glass
No
Reusable
Wide
Media Bottle