Learn More
Pyrex™ Class A Glass Cylinder with Polyethylene Stopper
Manufactured from chemically resistant Pyrex™ borosilicate glass.
Brand:  Pyrex 3212/16
 View more versions of this product
Product Code. 15737957
  • 769.00 € / Pack of 10
Estimated Shipment: 25-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Complies with ISO 4788
  • Printed with a batch code for downloadable batch certificate
  • Manufactured from chemically resistant Pyrex borosilicate glass
  • Graduations in white enamel for improved visibility with dark colored solutions
  • Supplied with polyethylene stopper
  • Robust hexagonal glass foot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Hexagonal
Glass
A
505 mm
45/40
Graduated Cylinder
ISO 4788
1000 mL
Polyethylene Stopper
Polyethylene
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Pyrex™ Class A Glass Cylinder with Polyethylene Stopper > Capacity: 1000mL; Graduations: 10mL