Learn More
Pyrex™ Class A Work Certified Glass Cylinder with Polyethylene Stopper
Manufactured from chemically resistant Pyrex™ borosilicate glass with hexagonal glass.
Brand:  Pyrex 3212/14WC
 View more versions of this product
Product Code. 15727957
  • 299.00 € / Pack of 2
Estimated Shipment: 25-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Complies with ISO 4788
  • Calibrated to Class A tolerances
  • Individually tested and supplied with certificate showing exact volume contained at 5 points on the scale
  • Conformity H certified
  • Printed with individual serial number for batch certification
  • Manufactured from chemically resistant Pyrex borosilicate glass with hexagonal glass
  • Graduations in white enamel for improved visibility with dark colored solutions
  • Supplied with polyethylene stopper
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Hexagonal
Glass
A
403 mm
34/35
Graduated Cylinder
ISO 4788
500 mL
Polyethylene Stopper
Polyethylene
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Pyrex™ Class A Work Certified Glass Cylinder with Polyethylene Stopper > Capacity: 500mL