missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ PYREX™ Cloning Cylinders
Create identical cells by surrounding a single cell or group of cells
59.79 € - 96.70 €
Specifications
Sterility | Non-sterile |
---|---|
Material | Borosilicate Glass |
Product Type | Cloning Cylinder |
Product Code | Brand | Diameter (English) | Diameter (Metric) | Height (English) | Height (Metric) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (English) | Diameter (Metric) | Height (English) | Height (Metric) | Price | Quantity & Availability | ||
12700668
|
Corning™
3166-6 |
0.23 in. | 6 mm | 0.31 in. | 8 mm |
59.79 €
Pack of 125 |
Estimated Shipment: 31-01-2025
Log in to see stock. |
||
11987595
|
Corning™
3166-8 |
0.31 in. | 8 mm | 0.31 in. | 8 mm |
69.00 €
Pack of 125 |
Estimated Shipment: 31-01-2025
Log in to see stock. |
||
11997595
|
Corning™
3166-10 |
0.39 in. | 10 mm | 0.39 in. | 10 mm |
96.70 €
Pack of 125 |
Estimated Shipment: 09-10-2024
Log in to see stock. |
||
Description
Clone a single cell or group of cells by surrounding them with this glass cylinder. Dip the end of the cylinder into a sterile silicone grease before pressing to the bottom of a culture flask to create an isolated well.
Specifications
Non-sterile | |
Borosilicate Glass | |
Cloning Cylinder |