Learn More
PYREX™ Culture Flask Closure
For use with 1135 culture flasks, 1138/10 Fernbach flasks and 1141 baffled flasks.
Brand:  Pyrex 4540/38P
63.22 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 12524917
  • 68.25 € / Pack of 10
Estimated Shipment: 19-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Two types of closure - Anodised aluminum - Two-position polypropylene
  • Suitable for autoclaving
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polypropylene
10
Autoclavable